Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helicobacter Pylori Antibody (HP/212) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311570
Description
Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, ImmunofluorescenceSpecifications
Helicobacter Pylori | |
Monoclonal | |
1 mg/ml | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
H. pylori | |
Total sonicate of Helicobacter pylori | |
0.1 mg | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Flow Cytometry, ELISA, Immunofluorescence, Immunofluorescence | |
HP/212 | |
Unconjugated | |
10 mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction