Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helicobacter Pylori Rabbit anti-Bacteria, Clone: RM429, Novus Biologicals™

Rabbit Monoclonal Antibody
$181.90 - $548.00
Specifications
| Antigen | Helicobacter Pylori |
|---|---|
| Clone | RM429 |
| Dilution | Immunohistochemistry 1:100-1:200 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
Description
Helicobacter Pylori Monoclonal specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Immunohistochemistry.Specifications
| Helicobacter Pylori | |
| Immunohistochemistry 1:100-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, 50% Glycerol, 1% BSA | |
| Proteins purified from Helicobacter pylori | |
| Primary | |
| Store at -20°C. |
| RM429 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Bacteria | |
| H. pylori | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title