Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Helicobacter pylori Urease B Monoclonal Antibody (GT962)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA516310
Description
Recommended positive controls: Helicobacter pylori. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
Specifications
Helicobacter pylori Urease B | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Mouse | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG2a |
Western Blot | |
GT962 | |
Unconjugated | |
H. pylori | |
Synthetic peptide corresponding to a region within amino acids 341 and 438 of Helicobacter pylori Urease B. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction