Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Helicobacter pylori Urease B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532168
Description
Recommended positive controls: Helicobacter pylori. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
Specifications
| Helicobacter pylori Urease B | |
| Polyclonal | |
| Unconjugated | |
| H. pylori | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of Helicobacter pylori Urease B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction