Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helios Antibody (22F6) - BSA Free, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP237723
Description
Helios Monoclonal specifically detects Helios in Human, Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Helios | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1 to 4 μg/mL, CyTOF-ready | |
HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
Armenian Hamster | |
Protein A purified | |
RUO | |
Primary | |
The armenian hamster monoclonal antibody 22F6 recognizes Helios, a transcription factor expressed in some hematopoietic stem cells, and at high levels in thymic-derived regulatory T cells. | |
Human, Mouse | |
Purified |
Flow Cytometry, CyTOF | |
22F6 | |
Unconjugated | |
PBS (pH 7.4) with 15mM Sodium Azide | |
IKZF2 | |
Peptide coresponding to the amino acids 51-107 of Helios. | |
0.1 mg | |
Adaptive Immunity, Immunology | |
22807 | |
Protein A purified | |
Store at 4C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction