Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helios Antibody (22F6), DyLight 550, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP237723R
Description
Helios Monoclonal specifically detects Helios in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Helios | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IKZF2 | |
Peptide coresponding to the amino acids 51-107 of Helios. | |
0.1 mL | |
Adaptive Immunity, Immunology | |
22807 | |
Human, Mouse | |
Purified |
Flow Cytometry | |
22F6 | |
Flow Cytometry | |
HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
Armenian Hamster | |
Protein A purified | |
RUO | |
Primary | |
The armenian hamster monoclonal antibody 22F6 recognizes Helios, a transcription factor expressed in some hematopoietic stem cells, and at high levels in thymic-derived regulatory T cells. | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction