Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helios Antibody (736440), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB73092A
Description
Helios Monoclonal antibody specifically detects Helios in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Helios | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
736440 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
E. coli-derived recombinant human Helios, Met1-Gln97, Accession # Q9UKS7 | |
0.1 mL | |
Adaptive Immunity, Immunology | |
22807 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction