Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helios Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204050AF700
Description
Helios Polyclonal antibody specifically detects Helios in Human samples. It is validated for Western BlotSpecifications
Helios | |
Polyclonal | |
Western Blot | |
HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human IKAROS Family Zinc Finger 2 (Helios) using the numbering given in entry NP_057344.2 (GeneID 22807). | |
0.1 mL | |
Adaptive Immunity, Immunology | |
22807 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction