Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Helix Elite™ Inactivated Standard SARS-CoV-2 B.1.1.7 Whole Virus (Alpha Variant)
Shop All Microbiologics Products

Click to view available options
Form:
Lyophilized
Quantity:
5 Pellets (Individually Packaged), 5 Vials of Molecular Standard Water
6 Swabs (Individually Packaged)
Description
- Fully intact, non-viable process controls
- Use for every step of the molecular testing process, including extraction, amplification, and detection of SARS-CoV-2
- Convenient, test-ready format
- Easy, economical room temperature storage
- Biosafety Level 1
Specifications
Specifications
| Format | Swabs |
| For Use With (Application) | Molecular Testing |
| Product Line | Helix Elite™ Inactivated Standard |
| Quantity | 6 Swabs (Individually Packaged) |
| Strain Designations | SARS-CoV-2 B.1.1.7 (Alpha Variant) Whole Virus |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction