Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Helix Elite™ Inactivated Standard SARS-CoV-2 B.1.351 Whole Virus (Beta Variant)
SDP

Catalog No. p-7235947 Shop All Microbiologics Products
Click to view available options
Form:
Lyophilized
Quantity:
6 Swabs (Individually Packaged)
This item is not returnable. View return policy
This item is not returnable. View return policy

The Inactivated SARS-CoV-2 B.1.351 Whole Virus (Beta Variant) can be used to monitor the extraction, amplification, and detection of SARS-CoV-2 molecular applications. Routine use of external quality controls monitor day to day performance of molecular testing procedures.

  • Fully intact, non-viable process controls
  • Use for every step of the molecular testing process, including extraction, amplification, and detection of SARS-CoV-2
  • Convenient, test-ready format
  • Easy, economical room temperature storage
  • Biosafety Level 1

Specifications

Format Swabs
For Use With (Application) Molecular Testing
Product Line Helix Elite™ Inactivated Standard
Quantity 6 Swabs (Individually Packaged)
Strain Designations SARS-CoV-2 B.1.351 (Beta Variant) Whole Virus
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.