Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HELLS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564099 Supplier Invitrogen™ Supplier No. PA564099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVYAPLSKKQ EIFYTAIVNR TIANMFGSSE KETIELSPTG RPKRRTRKSI NYSKIDDFPN ELEKLISQIQ PEVDRERAVV EVNIPV Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

This gene encodes a lymphoid-specific helicase. Other helicases function in processes involving DNA strand separation, including replication, repair, recombination, and transcription. This protein is thought to be involved with cellular proliferation and may play a role in leukemogenesis. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HELLS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HELLS
Gene Accession No. Q9NRZ9
Gene Alias AI323785; E130115I21Rik; helicase, lymphoid specific; helicase, lymphoid-specific; hells; ICF4; LSH; Lymphocyte-specific helicase; lymphoid-specific helicase; Lysh; Nbla10143; PASG; Proliferation-associated SNF2-like protein; SMARCA6; SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6; SWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, subfamily A, member 6; Tbc1d12; TBC1D12: TBC1 domain family, member 12; YFK8
Gene Symbols HELLS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HELLS. Recombinant protein control fragment (Product #RP-103497).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3070
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.