Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HELQ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588692 Supplier Invitrogen™ Supplier No. PA588692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T, LO2, HeLa, Raji, Mouse testis.

HEL308 is a single-stranded DNA-dependent ATPase and DNA helicase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HELQ
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Helq
Gene Accession No. Q2VPA6, Q8TDG4
Gene Alias AU021725; D430018E21Rik; DNA helicase Hel308; HEL308; Helicase POLQ-like; helicase, mus308-like; helicase, POLQ-like; HELQ; Mus308-like helicase; POLQ-like helicase
Gene Symbols Helq
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113510, 191578, 360912
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.