Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HELT Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981722
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1722 Supplier Bioss Supplier No. BS11851R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcriptional repressor which binds preferentially to the canonical E box sequence 5′-CACGCG-3′.

Specifications

Antigen HELT
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HELT
Gene Accession No. A6NFD8
Gene Alias A830086M02; bHLH transcription factor megane; bHLHb44; Hairy and enhancer of split-related protein HELT; HCM1228; Helt; helt bHLH transcription factor; Helt-ps1; HES/HEY-like transcription factor; HES/HEY-like transcription factor, pseudogene 1; Hesl; Heslike; Hey-like transcription factor; Hey-like transcriptional repressor; megane; Megane bHLH factor; mesencephalic GABAergic neurons enhancer of split related protein; Mgn; Protein Hes-like; Protein megane; RGD1564073
Gene Symbols HELT
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HELT.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 391723
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.