Learn More
HELZ Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA555812
Description
Immunogen sequence: TQVSQEVPEN CQEWIGGKMA QNGLDHYVYK VGIAFNTEIF GTFRQTIVFD FGLEPVLMQR VMIDAASTED LEYLMHAKQQ LVTTAKRWDS SSKTIIDFEP NETTDLEKSL LIRYQIPLSA DQLFTQSVLD KSLTKSNYQS RL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.

Specifications
HELZ | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P42694 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9931 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
HELZ | |
HELZ | |
Recombit protein corresponding to Human HELZ | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only