Learn More
Invitrogen™ HELZ2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total PRIC285 protein.
Specifications
Specifications
| Antigen | HELZ2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | HELZ2 |
| Gene Accession No. | Q9BYK8 |
| Gene Alias | ATP-dependent helicase PRIC285; helicase with zinc finger 2; helicase with zinc finger 2, transcriptional coactivator; Helicase with zinc finger domain 2; HELZ2; KIAA1769; PDIP1; PDIP-1; PDIP1-alpha; peroxisomal proliferator activated receptor A interacting complex 285; peroxisomal proliferator-activated receptor A interacting complex 285; peroxisomal proliferator-activated receptor A-interacting complex 285 kDa protein; peroxisomal proliferator-activated receptor alpha-interacting cofactor complex, 285 kD subunit; PPAR gamma DBD-interacting protein 1; PPAR-alpha-interacting complex protein 285; PPAR-gamma DBD-interacting protein 1; PPAR-gamma DNA-binding domain-interacting protein 1; PPARgamma-DNA-binding domain-interacting protein1; PPARG-DBD-interacting protein 1; PRIC complex; PRIC285 |
| Gene Symbols | HELZ2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.