Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HELZ2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101743 Supplier Invitrogen™ Supplier No. PA5101743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PRIC285.

The protein encoded by this gene is a nuclear transcriptional co-activator for peroxisome proliferator activated receptor alpha. The encoded protein contains a zinc finger and is a helicase that appears to be part of the peroxisome proliferator activated receptor alpha interacting complex. This gene is a member of the DNA2/NAM7 helicase gene family. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HELZ2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HELZ2
Gene Accession No. E9QAM5, Q9BYK8
Gene Alias ATP-dependent helicase PRIC285; helicase with zinc finger 2; helicase with zinc finger 2, transcriptional coactivator; Helicase with zinc finger domain 2; HELZ2; KIAA1769; PDIP1; PDIP-1; PDIP1-alpha; peroxisomal proliferator activated receptor A interacting complex 285; peroxisomal proliferator-activated receptor A interacting complex 285; peroxisomal proliferator-activated receptor A-interacting complex 285 kDa protein; peroxisomal proliferator-activated receptor alpha-interacting cofactor complex, 285 kD subunit; PPAR gamma DBD-interacting protein 1; PPAR-alpha-interacting complex protein 285; PPAR-gamma DBD-interacting protein 1; PPAR-gamma DNA-binding domain-interacting protein 1; PPARgamma-DNA-binding domain-interacting protein1; PPARG-DBD-interacting protein 1; PRIC complex; PRIC285
Gene Symbols HELZ2
Host Species Rabbit
Immunogen A synthesized peptide derived from human HELZ2(Accession Q9BYK8), corresponding to amino acid residues V2244-Y2294.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229003, 85441
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.