Learn More
MilliporeSigma Hemacolor™ Rapid Staining of Blood Smear
Shop All MilliporeSigma ProductsDescription
The Hemacolor™ Rapid staining of blood smear - staining set for microscopy, is used for human-medical cell diagnosis and serves the purpose of the hematological and clinico-cytological investigation of sample material of human origin, for example smears of whole blood and bone marrow.
The Hemacolor™ Rapid staining of blood smear staining set combines the brilliance and reproducibility of the Pappenheim staining with the time-advantage of a rapid stain. The Hemacolor™ Rapid staining of blood smear staining set contains a fixing solution, one red (eosin) and one blue (azur) staining solution and phosphate buffer tablets pH 7.2 according to Weise (Product number 1.09468). The staining solutions are separately applied, which results in a high degree of stability. The conventionally appearing precipitates in the hematological staining solutions, could be avoided in this kit. In addition to blood and bone marrow smears, Hemacolor™ Rapid staining of blood smear can be used in clinical cytology, e.g. for staining urine, sputum, FNAB, effusions and lavages.
A kit contains safe 500 mL glass bottles and six buffer tablets. The package is sufficient for 250 to 1000 applications. The buffer tablets according to Weise pH 7.2 (Product number 1.09468) are also available in larger pack sizes, thus, they can be useful for higher demands.
Specifications
Specifications
| Type | Hematology Stain |
| No. of Reactions | 250 to 1000 Applications |
| Product Type | Staining Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.