Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ HEMADO
GREENER_CHOICESDP

Catalog No. 157910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-791-0 10 mg
15-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-791-0 Supplier Tocris Bioscience™ Supplier No. 1579/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity selective A3 agonist

High affinity and selective adenosine A3 receptor agonist (Ki values are 1.1, 327, 1230 and > 30,000 nM for human A3, A1, A2A and A2B receptors respectively).

Chemical Identifiers

CAS 403842-38-6
Molecular Formula C17H23N5O4

Specifications

Quantity 10 mg
Formula Weight 361.4
Purity >99%
Molecular Weight (g/mol) 361.402
Chemical Name or Material HEMADO
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.