Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hemagglutinin-Neuraminidase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109232 Supplier Invitrogen™ Supplier No. PA5109232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Hemagglutinin-neuraminidase transfected HEK293 cell.

Hemagglutinin-neuraminidase refers to a single viral protein that has both hemagglutinin and neuraminidase activity. This is in contrast to the proteins found in influenza, where both functions exist but in two separate proteins. However it does show a structural similarity to influenza viral neuraminidase and has a six-bladed beta-propeller structure. Hemagglutinin-neuraminidase allows the virus to stick to a potential host cell, and cut itself loose if necessary. Hemagglutinin-neuraminidase can be found in a variety of paramyxoviruses including mumps virus, human parainfluenza virus 3, and the avian pathogen Newcastle disease virus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemagglutinin-Neuraminidase
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Host Species Rabbit
Immunogen Synthetic peptide within newcastle disease virus Hemagglutinin-neuraminidase aa 537-586.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.