Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hematin, Spectrum™ Chemical
SDP

Synonym: Hydroxyhemin, Ferrihemate, Protohematin, Ferriheme

Supplier:  Spectrum Chemical Manufacturing Corporation H1156250GMBL

Quantity 250 g

 View more versions of this product

Catalog No. 18-620-806


Only null left
Add to Cart
Request a Quote

Description

Description

Hematin is a porphyrin that contains a ferric iron ion with a chloride ligand. It is typically used to treat porphyria attacks, primarily in acute intermittent porphyria. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic uses.

Specifications

Chemical Identifiers

15489-90-4
633.5 g/mol

Specifications

15489-90-4
90% Min.
250 g
633.5 g/mol
Amber Glass Bottle
Hematin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.