Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hematin, Spectrum™ Chemical
SDP

Synonym: Hydroxyhemin, Ferrihemate, Protohematin, Ferriheme

$879.00 - $16490.00

Chemical Identifiers

CAS 15489-90-4
Molecular Weight (g/mol) 633.5 g/mol
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-620-804
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H11565GM
5 g
Each for $879.00
Only null left
Add to Cart
 
18-620-805
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H115625GM
25 g
Each for $2,261.00
Only null left
Add to Cart
 
18-620-806
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H1156250GMBL
250 g
Each for $16,490.00
Only null left
Add to Cart
 
Description

Description

Hematin is a porphyrin that contains a ferric iron ion with a chloride ligand. It is typically used to treat porphyria attacks, primarily in acute intermittent porphyria. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic uses.

Specifications

Chemical Identifiers

15489-90-4
633.5 g/mol

Specifications

15489-90-4
90% Min.
Hematin
633.5 g/mol
Amber Glass Bottle
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.