Learn More
Eisco™ Hematite Mineral Specimen
SCI_ED
Catalog No. S26619 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S26619 10/Pk.
S24693 1 Ea.
S24694 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S26619 Supplier Eisco™ Supplier No. ESNG0018PK10
Add to Cart
Add to Cart
Hematite Mineral Specimen is made up of iron and oxygen, giving it distinct red, earthy color.
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Visual aid for other iron-bearing rocks and minerals
Specifications
Product Type Mineral
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Hematite