Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Hematology Phosphate Buffer

Catalog No. p-4532411 Shop All Epredia Products
Click to view available options
pH:
6.6
7.2
Quantity:
9.25 g
12 x 9.25g
This item is not returnable. View return policy

Catalog No. 22-050-294

Supplier: Epredia™ 89032

This item is not returnable. View return policy

Combine Epredia™ Phosphate Buffer with Giemsa, Wright, and Wright-Giemsa stains for excellent results.

Supplied as 9.25g powder.

Specifications

Product Type Phosphate Buffered Saline (PBS)
Quantity 9.25 g
pH 6.6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.