Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hematopoietic Prostaglandin D Synthase/HPGDS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP154625
Description
Hematopoietic Prostaglandin D Synthase/HPGDS Polyclonal specifically detects Hematopoietic Prostaglandin D Synthase/HPGDS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Hematopoietic Prostaglandin D Synthase/HPGDS | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 2.5.1.18, EC 5.3.99.2, Glutathione S-transferase, glutathione S-transferase sigma, Glutathione-dependent PGD synthase, Glutathione-requiring prostaglandin D synthase, GST class-sigma, GSTShematopoietic prostaglandin D2 synthase, hematopoietic prostaglandin D synthase, H-PGDS, PGDSglutathione-dependent PGD synthetase, Prostaglandin-H2 D-isomerase, PTGDS2 | |
| Rabbit | |
| 23 kDa | |
| 100 μL | |
| Signal Transduction | |
| 27306 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| O60760 | |
| HPGDS | |
| Synthetic peptides corresponding to PGDS(prostaglandin D2 synthase, hematopoietic) The peptide sequence was selected from the N terminal of PGDS. Peptide sequence EQADWPEIKSTLPFGKIPILEVDGLTLHQSLAIARYLTKNTDLAGNTEME. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction