Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Hematoxylin
SDP

Catalog No. 522225G Shop All Novus Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-222-5G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 52-222-5G Supplier Tocris Bioscience™ Supplier No. 5222/25G

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Histologic stain; used to stain cell nuclei

Hematoxylin is a dye used to stain cell nuclei. Hematoxylin is commonly combined with eosin for H&E histological staining and is a component of the Papanicolaou stain used in cytology. Hematoxylin can be used with other fixatives to stain muscle striations, fibrin, myelin and elastin. Can also be used for H&E staining of intact tissues.

Specifications

For Use With (Application) Histology Stains
Molecular Formula (6 AS,11 bR)-7,11 b-Dihydrobenz[b]indeno[1,2-d]pyran-3,4,6 A,9,10(6 H)-pentol
CAS 517-28-2
Content And Storage Room Temperature
Quantity 25 g
Product Type Hematoxylin
Molecular Weight (g/mol) M.W. (Observed): 302.28
Inhibitors Hematoxylin

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.