Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hematoxylin, Spectrum™ Chemical
SDP

Catalog No. 18620894 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-894 5 g
18-620-895 10 g
18-620-897 100 g
18-620-898 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-894 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HE1155GM

Synonym: Haematoxylin, Hydroxybrazilin, Hydroxybrasilin, Natural Black 1

Hematoxylin is a a substance that is extracted from the heartwood of logwood tree and along with eosin stain is one of the most widely used stains in histology. It is a permanent stain as opposed to other stains that are only temporary in nature.

Chemical Identifiers

CAS 517-28-2

Specifications

Packaging Amber Glass
Color White to White/Yellow
Quantity 5 g
Chemical Name or Material Hematoxylin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.