Learn More
Invitrogen™ Heme oxygenase 2 Monoclonal Antibody (OTI1D10)

Description
Heme oxygenase 2 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | Heme oxygenase 2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1D10 |
| Concentration | 0.59 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HMOX2 |
| Gene Accession No. | P30519 |
| Gene Alias | heme oxygenase (decycling) 2; heme oxygenase 2; heme oxygenase-2 non-reducing isoform; HMOX 2; HMOX2; HO 2; HO2; HO-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.