Learn More
Invitrogen™ Heme oxygenase 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579378
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, human Caco-2 whole cell, human HEK293 whole cell, human Jurkat whole cell, human HepG2 whole cell, human K562 whole cell, human Raji whole cell. IHC: Human Lung Cancer Tissue.
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family. Several alternatively spliced transcript variants encoding three different isoforms have been found for this gene.
Specifications
Heme oxygenase 2 | |
Polyclonal | |
Unconjugated | |
HMOX2 | |
heme oxygenase (decycling) 2; heme oxygenase 2; heme oxygenase-2 non-reducing isoform; HMOX 2; HMOX2; HO 2; HO2; HO-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3163 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
P30519 | |
HMOX2 | |
E.coli-derived human HMOX2 recombinant protein (Position: S2-M316). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.