Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precipio™ HemeScreen CLL Limit of Detection Kit

Catalog No. 22021996
Change view
Click to view available options
Quantity:
5 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-996 5 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-021-996 Supplier Precipio™ Supplier No. HSLODCCLL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The HemeScreen CLL Limit of Detection Kit contains five control vials to verify the lower level of detection for the HemeScreen Cytopenia panel genes: MYD88, NOTCH1, CXCR4, and SF3B1.

The HemeScreen AML Limit of Detection Kit contains five control vials to verify the lower level of detection for the HemeScreen AML panel genes and mutant allelic frequency gene mutants at 1%, 3%, 5%, 10%, and 20%.

  • Genes: MYD88, NOTCH1, CXCR4, and SF3B1

Specifications

Content And Storage 6 x 1.5 mL Tube Mutant Controls. Store at -20°C
Control Template Mutant Control DNA
Detection Method HRM
Form Liquid
Format Kit, Tube(s)
PCR Method qPCR
Technique RT- PCR, HRM
Disease Chronic Lymphocytic Leukemia
For Use With (Equipment) PCR Instruments
Gene Symbol MYD88, NOTCH1, CXCR4, SF3B1
Genotyping Target MYD88, NOTCH1, CXCR4, SF3B1
Includes Label or Dye Yes
Kit Type HRM PCR Mutation Detection Kit
Label or Dye High Resolution Melting (HRM)
No. of Tests 5 Test
Primer PCR Primers
Product Line HemeScreen
Product Type Limit of Detection Kit
Quantity 5 Samples
Research Category Cancer, Hematology
Shipping Condition Cold Pack
Software HRM v3.1 Thermo Fisher Software
Sufficient For 5 Samples
Target MYD88, NOTCH1, CXCR4, SF3B1
Target Loci MYD88, NOTCH1, CXCR4, SF3B1
Target Organism Class Human
Compatibility PCR Instruments
Final Product Type Control DNA
Reaction Format Kit
Reagent Type Control Reagent
Sample Type Peripheral Blood, Bone Marrow
Starting Material 20 ng DNA
Includes 6 x 1.5 mL tubes mutant controls
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.