Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precipio™ HemeScreen IDH1 Kit-100

Catalog No. 22021979
Change view
Click to view available options
Quantity:
100 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-979 100 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-021-979 Supplier Precipio™ Supplier No. HSFFIDH1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The HemeScreen IDH1 Kit-100 is an RUO reagent set kit used to detect IDH1 gene mutations through HRM methods.

HemeScreen is a proprietary set of RUO (Research Use Only) reagents used to screen the wild type (Negative) from Mutated (Positive) genes in a simplified workflow relative to alternative molecular testing technologies (RT-PCR or NGS).

  • Simplified workflow and reduced chance of error
  • Contains primers mixed with master mix to run 100 samples in duplicate, and mutant controls to run 5 batches
  • Data capture during thermocycling
  • Fast turnaround time
  • Broad mutation detection (SNPs, Modifications, Indels)
  • Low limits of detection
  • Results are qualitative and DNA sequencing is required to identify specific isoforms
  • LOD: 5%
  • Assay includes Primers-Mastermix Mix and Positive controls
  • Requires HRM-enabled RT-PCR (example Thermo Fisher™ Quantstudio 3 or higher)

Specifications

Content And Storage 5 mL Tube Primer Set, 1.5 mL Tube Mutant Control. Store at -20°C
Control Template Mutant Positive Controls
Detection Method HRM
Form Liquid
Format Kit, Tube(s)
PCR Method qPCR
Technique PCR, HRM Analysis
For Use With (Equipment) PCR Instruments
Gene Symbol IDH1
Genotyping Target IDH1 Mutations
Includes Label or Dye Yes
Kit Type HRM IDH1 Mutation Detection Kit
Label or Dye High Resolution Melting (HRM)
No. of Tests 100 Test
Primer PCR Primers
Product Line HemeScreen
Product Type Genotyping Kit (HRM)
Quantity 100 Samples
Research Category Cancer Research, Hematology, Genetics
Sensitivity 0.95
Shipping Condition Dry Ice
Sufficient For 100 Samples
Target IDH1
Target Loci IDH1
Target Organism Class Human
Target Specificity >99%
Compatibility HRM-enabled RT- PCR Instruments, RT- PCR System
Final Product Type Mutation Data
No. of Reactions 100 Reactions
Reaction Format Master Mix
Reagent Type Genotyping Reagent
Sample Type Whole Blood, Bone Marrow
Starting Material DNA
Volume 20 μL
Includes 5 mL tube primer set, 1.5 mL tube mutant control
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.