Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precipio™ HemeScreen JAK2 Exon 14 Kit-100

Catalog No. 22021974
Change view
Click to view available options
Quantity:
100 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-974 100 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-021-974 Supplier Precipio™ Supplier No. HSFFJ14
Add to Cart
Add to Cart
This item is not returnable. View return policy

The HemeScreen JAK2 Exon 14 Kit-100 is an RUO reagent set kit used to detect JAK2 Exon 14 (V617F) gene mutations through HRM methods.

HemeScreen is a proprietary set of RUO (Research Use Only) reagents used to screen the wild type (Negative) from Mutated (Positive) genes in a simplified workflow relative to alternative molecular testing technologies (RT-PCR or NGS).

  • Simplified workflow and reduced chance of error
  • Contains primers mixed with master mix to run 100 samples in duplicate, and mutant controls to run 5 batches
  • Data capture during thermocycling
  • Fast turnaround time
  • Broad mutation detection (SNPs, Modifications, Indels)
  • Low limits of detection
  • Results are qualitative and DNA sequencing is required to identify specific isoforms
  • LOD: 5%
  • Assay includes Primers-Mastermix Mix and Positive controls
  • Requires HRM-enabled RT-PCR (example Thermo Fisher™ Quantstudio 3 or higher)

Specifications

Content And Storage 5 mL Tube Primer Set, 1.5 mL Tube Mutant Control. Store at -20°C
Detection Method HRM
Format Kit
PCR Method qPCR
Technique HRM, PCR
Disease Myeloproliferative Neoplasms
For Use With (Equipment) PCR Instruments
Gene Symbol JAK2
Genotyping Target JAK2 V617F
Includes Label or Dye Yes
Kit Type JAK2 Mutation Detection Kit
Label or Dye High Resolution Melting (HRM)
No. of Tests 100 Test
Primer PCR Primers
Product Line HemeScreen
Product Type Genotyping Kit
Quantity 100 Samples
Research Category Cancer, Hematology
Sensitivity 0.95
Shipping Condition Dry Ice
Sufficient For 100 Samples
Target JAK2 Exon 14
Target Loci JAK2 (Exon 14)
Target Organism Class Human
Target Specificity >99%
For Use With (Application) Genotyping
Compatibility HRM-enabled RT-PCR Instruments, RT-PCR, NGS
Reaction Format Master Mix
Reagent Type Genotyping Reagent
Starting Material DNA
Includes 5 mL tube primer set, 1.5 mL tube mutant control
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.