Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precipio™ HemeScreen MPN Limit of Detection Kit

Catalog No. 22021994
Change view
Click to view available options
Quantity:
5 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-994 5 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-021-994 Supplier Precipio™ Supplier No. HSLODCMPN
Add to Cart
Add to Cart
This item is not returnable. View return policy

The HemeScreen MPN Limit of Detection Kit contains five control vials to verify the lower levels of detection for the HemeScreen MPN panel genes: JAK2 Exon 12, JAK2 Exon 13, JAK2 Exon14 (V617F), CALR, and MPL.

The HemeScreen AML Limit of Detection Kit contains five control vials to verify the lower level of detection for the HemeScreen AML panel genes and mutant allelic frequency gene mutants at 1%, 3%, 5%, 10%, and 20%.

  • Genes: JAK2 Exon 12, JAK2 Exon 13, JAK2 Exon14 (V617F), CALR, and MPL

Specifications

Content And Storage 6 x 1.5 mL Tube Mutant Controls. Store at -20°C
Control Template Mutant Control
Detection Method HRM
Form Liquid
Format Kit, Tube(s)
PCR Method Standard PCR
Technique PCR, HRM
Disease Myeloproliferative Neoplasms
For Use With (Equipment) PCR Instruments
Gene Symbol JAK2, CALR, MPL
Genotyping Target JAK2 Exon 12, JAK2 Exon 13, JAK2 Exon 14 (V617F), CALR, MPL
Includes Label or Dye Yes
Kit Type MPN Gene Detection Kit
Label or Dye High Resolution Melting (HRM)
No. of Tests 5 Test
Product Line HemeScreen
Product Type Genotyping Kit
Quantity 5 Samples
Research Category Hematology, Cancer Research
Shipping Condition Cold Pack
Sufficient For 5 Samples
Target JAK2 Exon 12, JAK2 Exon 13, JAK2 Exon14 (V617F), CALR, MPL
Target Organism Class Human
For Use With (Application) Genotyping
Compatibility PCR Instruments
Reagent Type Genotyping Control
Includes 6 x 1.5 mL tube mutant controls
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.