Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precipio™ HemeScreen MPN Panel DNA Validation Set

Catalog No. 22021997
Change view
Click to view available options
Quantity:
20 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-997 20 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-021-997 Supplier Precipio™ Supplier No. HSDNACMPN
Add to Cart
Add to Cart
This item is not returnable. View return policy

The HemeScreen MPN Panel DNA Validation Kit contains 20 blinded control samples and pre-plated MPN panels.

Results are verified with a key for panel accuracy, sample positive percent agreement (PPA), and negative percent agreement (NPA).

  • 20 blinded control samples with at least 3 mutant and 3 normal alleles for each gene
  • 1 pre-plated, 4-sample MPN panel plate
  • 2 pre-plated, 8-sample MPN panel plates

Specifications

Content And Storage 20 x 1.5 mL Tubes Extracted DNA, 2 x 8-sample Plates, 4-sample Plate. Store at -20°C
Detection Method HRM
For Use With (Equipment) PCR Instruments
Includes Label or Dye Yes
Quantity 20 Samples
Shipping Condition Dry Ice
Target MPN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.