Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precipio™ HemeScreen MPN Panels

Catalog No. 22021969
Change view
Click to view available options
Quantity:
4 Samples
8 Samples
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-969 8 Samples
22-021-968 4 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-021-969 Supplier Precipio™ Supplier No. HS8PMPN
Add to Cart
Add to Cart
This item is not returnable. View return policy

The HemeScreen MPN Panel-4 is an RUO pre-plated kit used to detect MPN-related gene mutations for JAK2, MPL, and CALR genes through HRM methods.

HemeScreen is a proprietary set of RUO (Research Use Only) reagents used to screen the wild type (Negative) from Mutated (Positive) genes in a simplified workflow relative to alternative molecular testing technologies (RT-PCR or NGS).

  • Pre-plated configuration with embedded mutant, wild type, and NTC controls to run 4 or 8 samples in duplicate
  • Simplified workflow and reduced chance of error
  • Data capture during thermocycling
  • Fast turnaround time
  • Broad mutation detection (SNPs, Modifications, Indels)
  • Low limits of detection
  • Results are qualitative and DNA sequencing is required to identify specific isoforms
  • LOD: 5%
  • Assay includes Primers-Mastermix Mix and controls
  • Requires HRM-enabled RT-PCR (example Thermo Fisher™ Quantstudio 3 or higher)

Specifications

Content And Storage 96-well Plate with Gene Targets, Positive, Negative, and Wild Type Controls. Store at -20°C
Detection Method HRM
Form Liquid
Format Kit, 96-well Plate
PCR Method qPCR
Technique PCR, HRM Analysis
Disease Myeloproliferative Neoplasms
For Use With (Equipment) PCR Instruments
Gene Symbol JAK2,MPL,CALR
Genotyping Target JAK2, MPL, CALR
Includes Label or Dye Yes
Kit Type HRM DNA Detection Kit
Label or Dye High Resolution Melting (HRM)
No. of Tests 8 Test
Primer PCR Primers
Product Line HemeScreen
Product Type PCR Assay Kit
Quantity 8 Samples
Research Category Cancer, Genetics
Sensitivity 0.95
Shipping Condition Dry Ice
Sufficient For 8 Samples
Target JAK2 Ex. 14 (V617F), JAK2 Ex. 12, JAK2 Ex. 13, MPL, CALR
Target Loci JAK2 (Exon 12, Exon 13, Exon 14), MPL, CALR
Target Organism Class Human
Target Specificity >99%
For Use With (Application) Genotyping
Compatibility HRM-enabled RT-PCR Instruments
Reaction Format Master Mix
Reagent Type PCR Reagent, HRM Reagent
Sample Type Whole Blood, Bone Marrow
Starting Material DNA
Volume 20 μL
Includes 96-well plate with primers and mastermix, positive controls, negative controls (NTC), wild-type controls
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.