Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precipio™ HemeScreen WT1 Kit-100

Catalog No. 22021988
Change view
Click to view available options
Quantity:
100 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-988 100 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-021-988 Supplier Precipio™ Supplier No. HSFFWT1
Add to Cart
Add to Cart
This item is not returnable. View return policy

The HemeScreen WT1 Kit-100 is an RUO reagent set kit used to detect WT1 gene mutations through HRM methods.

HemeScreen is a proprietary set of RUO (Research Use Only) reagents used to screen the wild type (Negative) from Mutated (Positive) genes in a simplified workflow relative to alternative molecular testing technologies (RT-PCR or NGS).

  • Simplified workflow and reduced chance of error
  • Contains primers mixed with master mix to run 100 samples in duplicate, and mutant controls to run 5 batches
  • Data capture during thermocycling
  • Fast turnaround time
  • Broad mutation detection (SNPs, Modifications, Indels)
  • Low limits of detection
  • Results are qualitative and DNA sequencing is required to identify specific isoforms
  • LOD: 5%
  • Assay includes Primers-Mastermix Mix and Positive controls
  • Requires HRM-enabled RT-PCR (example Thermo Fisher™ Quantstudio 3 or higher)

Specifications

Content And Storage 5 mL Tube Primer Set, 1.5 mL Tube Mutant Control. Store at -20°C
Control Template Mutant Control
Detection Method HRM
Form Liquid
Format Kit, Tube(s)
PCR Method qPCR
For Use With (Equipment) PCR Instruments
Gene Symbol WT1
Genotyping Target WT1 Mutations
Includes Label or Dye Yes
Kit Type WT1 Mutation Detection Kit
Label or Dye High Resolution Melting (HRM)
No. of Tests 100 Test
Optimal Reaction Temperature 68°C
Primer PCR Primers
Product Line HemeScreen
Product Type Genotyping Kit (HRM)
Quantity 100 Samples
Research Category Cancer Research, Hematology, Molecular Biology
Sensitivity 0.95
Shipping Condition Dry Ice
Sufficient For 100 Samples
Target WT1
Target Loci WT1
Target Specificity >99%
Activation Time 3 min.
Compatibility HRM-enabled RT-PCR Instruments
Reaction Format Master Mix
Sample Type DNA from Whole Blood, DNA from Bone Marrow
Starting Material DNA
Volume 20 μL
Includes 5 mL tube primer set, 1.5 mL tube mutant control
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.