Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEMGN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554036 Supplier Invitrogen™ Supplier No. PA554036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VISVIQDHPF KMYQDMAKRE DLAPKMCQEA AVPKILPCPT SEDTADLAGC SLQAYPKPDV PKGYILDTDQ NPAEPEEYNE TDQGI Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 34%.

HEMGN regulates the proliferation and differentiation of hematopoietic cells. Overexpression of HEMGN block the TPA-induced megakaryocytic differentiation in the K562 cell model. HEMGN may also prevent cell apoptosis through the activation of the nuclear factor-kappa B (NF-kB).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEMGN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEMGN
Gene Accession No. Q9BXL5
Gene Alias 4921524M03Rik; AI317176; CT155; EDAG; EDAG-1; erythroid differentiation-associated gene protein; Hemgn; Hemogen; hemopoietic gene protein; Hgn; mNDR; NDR; negative differentiation regulator protein; nuclear protein; PRO1037; PRO1620; Unknown (protein for MGC:134079)
Gene Symbols HEMGN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HEMGN. Recombinant protein control fragment (Product #RP-92300).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55363
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.