Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HEMK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557023
Description
Immunogen sequence: VSNPPYVFHQ DMEQLAPEIR SYEDPAALDG GEEGMDIITH ILALAPRLLK DSGSIFLEVD PRHPELVSSW LQSRPDLYLN LVAVRRDFCG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
N5-glutamine methyltransferase responsible for the methylation of the glutamine residue in the universally conserved GGQ motif of the mitochondrial translation release factor MTRF1L. [UniProt].
Specifications
HEMK1 | |
Polyclonal | |
Unconjugated | |
HEMK1 | |
2310008M14Rik; AW049265; HEMK; HEMK homolog 7kb; hemK methyltransferase family member 1; HEMK1; M.HsaHemKP; MTQ1; MTRF1L release factor glutamine methyltransferase; RGD1308293; testis secretory sperm-binding protein Li 225n | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51409 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5R4 | |
HEMK1 | |
Recombinant protein corresponding to Human HEMK1. Recombinant protein control fragment (Product #RP-96312). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction