Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEMK1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983781
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3781 Supplier Bioss Supplier No. BS15452R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probable protein methyltransferase. May methylate a Gln residue in target proteins.

Specifications

Antigen HEMK1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HEMK1
Gene Accession No. Q9Y5R4
Gene Alias HEMK; HEMK homolog 7kb; HemK methyltransferase family member 1; HEMK1; M.HsaHemKP; MTQ1; MTRF1L release factor glutamine methyltransferase; testis secretory sperm-binding protein Li 225n
Gene Symbols HEMK1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HEMK1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51409
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.