Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEMK2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983782
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3782 Supplier Bioss Supplier No. BS15453R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Heterodimeric methyltransferase that catalyzes N5-methylation of ETF1 on ′Gln-185′, using S-adenosyl L-methionine as methyl donor. ETF1 needs to be complexed to ERF3 in its GTP-bound form to be efficiently methylated. May play a role in the modulation of arsenic-induced toxicity. May be involved in the conversion of monomethylarsonous acid (3+) into the less toxic dimethylarsonic acid.

Specifications

Antigen HEMK2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene N6AMT1
Gene Accession No. Q9Y5N5
Gene Alias 5830445C04Rik; BOS_547; C21orf127; HemK methyltransferase family member 2; HEMK2; LOC100352597; LOC615200; M.HsaHemK2P; Methylarsonite methyltransferase N6AMT1; Methyltransferase N6AMT1; M-N6DNA MTase A; MTQ2; N(6)-adenine-specific DNA methyltransferase 1; N(6)-adenine-specific DNA methyltransferase 1-like; N-6 adenine-specific DNA methyltransferase 1; N-6 adenine-specific DNA methyltransferase 1 (putative); N6AMT; N6AMT1; N6-DNA methyltransferase A; N6-DNA-methyltransferase; Pred28; Protein N(5)-glutamine methyltransferase; RGD1311843
Gene Symbols N6AMT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HEMK2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29104
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.