Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HemoCue America AlbuTrol™ for Albumin 201 System

Catalog No. 22601015
Click to view available options
Concentration:
22 to 28 mg/mL
70 to 80 mg/mL
Level:
High
Low
This item is not returnable. View return policy
This item is not returnable. View return policy

For use in verification of precision/accuracy for HemoCue Albumin 201 system

  • Assayed albumin control intended for professional use
  • Custom formulation which mimics fresh human urine
  • Available in two physiologic levels
  • Plastic dropper bottle (1mL) with dropper tip and colored screw cap
  • Shelf life is 15 months at 2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Content And Storage 2°C to 8°C
Concentration 22 to 28 mg/mL
For Use With (Equipment) HemoCue™ Albumin 201
Type AlbuTrol™ Albumin Control
Includes Dropper tip and colored screw cap
Intended Use For use in verification of precision/accuracy
Level Low
Quantity 2 x 1 mL
Source Human Urine Containing Albumin
Stability 1 Month
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.