Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HemoCue America AlbuTrol™ for Albumin 201 System
Click to view available options
Concentration:
22 to 28 mg/mL
70 to 80 mg/mL
Level:
High
Low
Description
- Assayed albumin control intended for professional use
- Custom formulation which mimics fresh human urine
- Available in two physiologic levels
- Plastic dropper bottle (1mL) with dropper tip and colored screw cap
- Shelf life is 15 months at 2° to 8°C

Specifications
Specifications
Color | Clear |
Content And Storage | 2°C to 8°C |
Concentration | 22 to 28 mg/mL |
For Use With (Equipment) | HemoCue™ Albumin 201 |
Type | AlbuTrol™ Albumin Control |
Includes | Dropper tip and colored screw cap |
Intended Use | For use in verification of precision/accuracy |
Level | Low |
Quantity | 2 x 1 mL |
Source | Human Urine Containing Albumin |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction