Learn More
HemoCue America Glucose Control Solution
Description
- Plastic dropper bottle (2mL) with colored screw cap
- Shelf life is 13 months at 2° to 8°C, after opening 30 days at 2° to 30°C°Delivered with whole blood and plasma equivalent values at physiologically relevant glucose values
- Conveniently packaged with one vial of each level per barcoded box
Specifications
Specifications
| Certifications/Compliance | IVD, CE |
| Color | Clear |
| Content And Storage | 2°C to 8°C |
| Format | Plastic Vial |
| For Use With (Equipment) | HemoCue™ B-Glucose and HemoCue™ Glucose 201 system |
| Type | GlucoTrol™-AQ Glucose Control Solution |
| Includes | Water, Glucose, Buffering salts |
| Intended Use | For professional use in the verification of the precision and accuracy of the HemoCue™ B-Glucose analyzer |
| Level | 1 Low, 3 High |
| pH | 7 to 7.5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.