Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HemoCue America Hb 201+ Analyzer: Cuvettes
Click to view available options
Quantity:
100
200
Description
Break-resistant, disposable cuvette collects the exact amount of blood and mixes the sample with the reagents automatically.

Specifications
Specifications
Product Type | Cuvettes for HB-201 Analyzer |
Sample Size | 10 μL |
Operating Temperature Range (English) | 59°F to 86°F |
Operating Temperature Range (Metric) | 15°C to 30°C |
For Use With (Equipment) | Hb 201 analyzer |
Quantity | 100 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction