Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HemoCue America HemoCue™ Hb 801 System

Catalog No. 22601036
Change view
Click to view available options
Analysis Time:
<1 sec.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Analysis Time
22-601-036 <1 sec.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-601-036 Supplier HemoCue America Supplier No. 121920
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HemoCue™ Hb 801 System is intended for the quantitative determination of hemoglobin in capillary or venous whole blood (K2EDTA and Li-Heparin) in point-of-care settings.

  • Wireless connectivity through middleware
  • Test results in under one second
  • Reagent-free cuvettes improve open-vial stability
  • Large display and intuitive on-screen symbols
  • Perform internal quality control self-tests on the analyzer (Refer to local or regulatory agency for any external control requirements)
  • CLIA-waived
  • HemoCue OnCue™ Support:
    • Extensive in-field support from a HemoCue representative
    • Comprehensive on-site or online product training programs
    • Live customer service and technical support when needed
  • Factory calibrated against the ICSH reference method, needs no further calibration
  • Measuring range 1.0 to 25.6 g/dL
  • Obtain results in 3 simple steps: fill microcuvette, place microcuvette into analyzer, and view results.
  • Operating Manual # 901912 190204 US
  • HemoCue Hb 801 Microcuvettes are sold separately
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Capillary and Venous Whole Blood
Analysis Time <1 sec.
Sample Size 10 μL
Includes HemoCue™ Hb 801 Analyzer, USB adapter. (Microcuvettes are sold separately.)
Width (English) 5.6 in.
Weight (English) 0.55 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.