Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hemogen Antibody (OTI2D7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272005B
Description
Hemogen Monoclonal specifically detects Hemogen in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Hemogen | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
HEMGN | |
Recombinant protein fragment corresponding to amino acids 260-484 of human Hemogen (NP_060907) produced in E.coli. | |
0.1 mL | |
55363.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2D7 | |
Western Blot, Immunohistochemistry | |
EDAGEDAG-1, Erythroid differentiation-associated gene protein, hemogen, Hemopoietic gene protein, NDR, Negative differentiation regulator protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction