Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemogen/EDAG Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982121
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2121 Supplier Bioss Supplier No. BS12303R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HEMGN regulates the proliferation and differentiation of hematopoietic cells. Overexpression of HEMGN block the TPA-induced megakaryocytic differentiation in the K562 cell model. HEMGN may also prevent cell apoptosis through the activation of the nuclear factor-kappa B (NF-kB).

Specifications

Antigen Hemogen/EDAG
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HEMGN
Gene Accession No. Q9BXL5
Gene Alias 4921524M03Rik; AI317176; BOS_8980; CT155; EDAG; EDAG-1; Erythroid differentiation-associated gene protein; Hemgn; Hemogen; Hemopoietic gene protein; Hgn; mNDR; NDR; Negative differentiation regulator protein; nuclear protein; PRO1037; PRO1620
Gene Symbols HEMGN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Hemogen/EDAG.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55363
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.