Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hemoglobin A1c Antibody (75C9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP232769
Description
Hemoglobin A1c Monoclonal specifically detects Hemoglobin A1c in Human samples. It is validated for Immunoassay.Specifications
| Hemoglobin A1c | |
| Monoclonal | |
| Unconjugated | |
| alpha one globin, alpha-1 globin, alpha-1-globin, alpha-globin, CD31, hemoglobin alpha 1 globin chain, Hemoglobin alpha chain, hemoglobin alpha-1 chain, hemoglobin subunit alpha, hemoglobin, alpha 1, MGC126895, MGC126897 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Native human HbA1c, does not cross-react with non-glycated hemoglobin. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| Immunoassay | |
| 75C9 | |
| Immunoassay | |
| HBA1 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with glycated (in vitro) synthetic peptide corresponding to the N-terminal end of beta-globin. | |
| 0.2 mg | |
| Signal Transduction | |
| 3039 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction