Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hemoglobin alpha Antibody, Alexa Fluor™ 647 , Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB11041083AF647
Description
Hemoglobin alpha Polyclonal antibody specifically detects Hemoglobin alpha in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Hemoglobin alpha | |
Polyclonal | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
Alexa Fluor 647 | |
alpha one globin, alpha-1 globin, alpha-1-globin, alpha-globin, CD31, hemoglobin alpha 1 globin chain, Hemoglobin alpha chain, hemoglobin alpha-1 chain, hemoglobin subunit alpha, hemoglobin, alpha 1, MGC126895, MGC126897 | |
Human Hemoglobin | |
1 mL | |
Signal Transduction | |
3039 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction