Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hemoglobin Antibody (2A1), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320120MFV500
Description
Hemoglobin Monoclonal antibody specifically detects Hemoglobin in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
Hemoglobin | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Mouse | |
Purified |
Western Blot, ELISA, Immunocytochemistry | |
2A1 | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
alpha one globin;alpha-1 globin;alpha-1-globin;alpha-2 globin chain;Alpha-globin;ALPHA-GLOBIN GENE;delta globin;Haemoglobin;Hb;HbA1c;hemoglobin alpha 1 globin chain;Hemoglobin alpha chain;Hemoglobin subunit alpha;hemoglobin, alpha 1 | |
Murine hybridomas were generated using spleen cells from aging (8-10 month old) NZB/W F1 animals. Hybridomas secreting antibodies reactive to murine hemoglobin were subcloned by limiting dilution and antibody isotypes determined using commercial reagents. Eight-week-old NZB/W F1 were immunized subcutaneously with 50 mg mHb antigen emulsified in Complete Fruend's Adjuvant. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction