Learn More
Hemoglobin beta A-2 Monoclonal Antibody (21G1.F1.B9.G9.D11), Rockland™

Mouse Monoclonal Antibody
$174.73 - $651.91
Specifications
Antigen | Hemoglobin beta A-2 |
---|---|
Clone | 21G1.F1.B9.G9.D11 |
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0627
![]() ![]() |
Rockland Immunochemicals
200301GS8 |
100 μg |
Each for $651.91
|
|
|||||
50-199-0628
![]() ![]() |
Rockland Immunochemicals
200301GS8S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Reacts specifically with human HbA-2 delta isoform. This antibody does not react with the HbA, HbS, HbC, or HbF isoforms.
Hemoglobin subunit delta is a protein that in humans is encoded by the HBD gene. The delta (HBD) and beta (HBB) genes are normally expressed in the adult: two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin. Two alpha chains plus two delta chains constitute HbA-2, which with HbF comprises the remaining 3% of adult hemoglobin. Five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5'-epsilon-Ggamma-Agamma-delta-beta-3'.Specifications
Hemoglobin beta A-2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
delta globin; Delta-globin; delta-globin chain; HBD; Hemoglobin delta chain; Hemoglobin subunit delta; hemoglobin, delta | |
HBD | |
IgG1 κ | |
-20°C, Avoid Freeze/Thaw Cycles | |
HBD |
21G1.F1.B9.G9.D11 | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P02042 | |
100187828, 3045 | |
Anti-Hemoglobin beta A-2 Monoclonal Antibody was produced in mice by repeated immunizations with synthetic peptide corresponding to amino acid residues near the N-terminus of Hb delta-subunit conjugated to KLH. | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.